5-Acetyl-2,3-dihydrobenzo[b]furan ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A124581-1G A124581-250MG A124581-25G A124581-5G
ALA124581-1GEA 19.86 USD
ALA124581-1G ALA124581250MG ALA124581-25G ALA124581-5G
5-Acetyl-2,3-dihydrobenzo[b]furan ≥97%
5-Acetyl-2,3-dihydrobenzo[b]furan
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 120-121°C
Melting Pt: 56-59°C
MDL Number: MFCD00068033
CAS Number: 90843-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ddca0efba285d4be14a1e2ebf6343d4178a4cab&vid=7ddca0efba285d4be14a1e2ebf6343d4178a4cab&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR