3-(Piperidin-2-yl)pyridine ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
(±) Anabasine, 2-Pyridin-3-ylpiperidine
A124541-100MG
A124541-1G
A124541-250MG
A124541-25MG
A124541-5G
ALA124541100MGEA
114.86
USD
ALA124541100MG
ALA124541-1G
ALA124541250MG
ALA124541-25MG
ALA124541-5G
3-(Piperidin-2-yl)pyridine ≥97%
3-(Piperidin-2-yl)pyridine
Anabasine inhibited androstenedione conversion to estrogen in a dose-dependent mannera tobacco alkaloid, was used as a biomarker of active tobacco use
Formula:
C₁₀H₁₄N₂ MW: 162.236 g/mol Boiling Pt: 270-272°C Melting Pt: 9°C Density: 1.046 g/cm³ Flash Pt: 93°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006370 CAS Number: 13078-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...