4-Bromo-1-naphthylamine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-Amino-4-bromonaphthalene

A124442-100G A124442-25G A124442-5G
ALA124442-100GEA 601.26 USD
ALA124442-100G ALA124442-25G ALA124442-5G
4-Bromo-1-naphthylamine ≥98%
4-Bromo-1-naphthylamine
Formula: C₁₀H₈BrN
MW: 222.08 g/mol
Melting Pt: 100.0 to 104.0 °C
Storage Temperature: Ambient
MDL Number: MFCD00004023
CAS Number: 2298-07-9
EINECS: 218-944-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR