(R)-2-Aminobutanamide hydrochloride ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (R)-(-)-2-Aminobutanamide HCl, (R)-2-Aminobutyramide hydrochloride, (2R)-2-Aminobutanamide Hydrochloride, (R)-(–)-2-Aminobutanamide hydrochloride

A124384-1G A124384-25G A124384-5G
ALA124384-1GEA 61.66 USD
ALA124384-1G ALA124384-25G ALA124384-5G
(R)-2-Aminobutanamide hydrochloride ≥96%
(R)-2-Aminobutanamide hydrochloride

Product Application:R)-2-Aminobutyramide Hydrochloride is used as a reagent in the synthesis of a new class of 2-[(arylalkyl)amino]alkanamide derivatives which have anticonvulsant activity.

Formula: C₄H₁₁ClN₂O
MW: 138.59 g/mol
Melting Pt: 258-262°C
Storage Temperature: Ambient
MDL Number: MFCD09265126
CAS Number: 103765-03-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d53831f36f93dbac4bd0feb15c5414e6b9096a8&vid=9d53831f36f93dbac4bd0feb15c5414e6b9096a8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR