1,3,5-Adamantanetriol ≥95%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
Adamantane-1,3,5-triol, 1,3,5-Trihydroxyadamantane
A124366-1G
A124366-250MG
ALA124366-1GEA
130.06
USD
ALA124366-1G
ALA124366250MG
1,3,5-Adamantanetriol ≥95%
1,3,5-Adamantanetriol
Formula:
C₁₀H₁₆O₃ MW: 184.24 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08276262 CAS Number: 99181-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...