2-Amino-3-(trifluoromethyl)pyridine ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A124359-100G
A124359-1G
A124359-25G
A124359-5G
ALA124359-100GEA
3507.31
USD
ALA124359-100G
ALA124359-1G
ALA124359-25G
ALA124359-5G
2-Amino-3-(trifluoromethyl)pyridine ≥97%
2-Amino-3-(trifluoromethyl)pyridine
Formula:
C₆H₅F₃N₂ Melting Pt: 70-72°C |
MDL Number:
MFCD04972538 CAS Number: 183610-70-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...