2-Amino-6-(trifluoromethyl)pyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A124358-100G A124358-1G A124358-250MG A124358-25G A124358-5G
ALA124358-100GEA 994.56 USD
ALA124358-100G ALA124358-1G ALA124358250MG ALA124358-25G ALA124358-5G
2-Amino-6-(trifluoromethyl)pyridine ≥98%
2-Amino-6-(trifluoromethyl)pyridine

Substrate used in a one-pot synthesis of the corresponding 7-aza-indole. Substrate used in a one-pot synthesis of the corresponding 7-aza-indole.

Formula: C₆H₅F₃N₂
Melting Pt: 86-89°C
MDL Number: MFCD03093868
CAS Number: 34486-24-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a63c99ce1f2e6d09d97878a908b1765a172dba1a&vid=a63c99ce1f2e6d09d97878a908b1765a172dba1a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR