2-Amino-4-(trifluoromethyl)pyridine ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A124356-1G A124356-250MG A124356-25G A124356-5G A124356-100G A124356-10G
ALA124356-1GEA 36.96 USD
ALA124356-1G ALA124356250MG ALA124356-25G ALA124356-5G ALA124356-100G ALA124356-10G
2-Amino-4-(trifluoromethyl)pyridine ≥99%
2-Amino-4-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
Melting Pt: 71-74°C
MDL Number: MFCD00077455
CAS Number: 106447-97-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bf25b6dbb3ab3182781d864bfc1f021b38f9681&vid=3bf25b6dbb3ab3182781d864bfc1f021b38f9681&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR