1,5-Dihydroxy-9,10-anthraquinone 85

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1,5-Dihydroxyanthraquinone, Anthrarufin

A124340-100G A124340-10G A124340-25G A124340-5G
ALA124340-100GEA 81.61 USD
ALA124340-100G ALA124340-10G ALA124340-25G ALA124340-5G
1,5-Dihydroxy-9,10-anthraquinone 85
1,5-Dihydroxy-9,10-anthraquinone
Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Melting Pt: 279 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001210
CAS Number: 117-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d9cd110fbfc9a4f52ec1d5bf2fdbb006e2c4f71&vid=9d9cd110fbfc9a4f52ec1d5bf2fdbb006e2c4f71&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR