2-Amino-5-bromo-4-methylpyridine ≥98%

Supplier: Aladdin Scientific
Warning

A124300-25G A124300-5G
ALA124300-25GEA 175.66 USD
ALA124300-25G ALA124300-5G
2-Amino-5-bromo-4-methylpyridine ≥98%
2-Amino-5-bromo-4-methylpyridine
Formula: C₆H₇BrN₂
Melting Pt: 148-151°C
MDL Number: MFCD03427660
CAS Number: 98198-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1f7df6317102dcf4ac7e57832e0e1d25a244199&vid=b1f7df6317102dcf4ac7e57832e0e1d25a244199&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR