6-Nitro-o-anisidine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A124291-100G A124291-10G A124291-1G A124291-250MG A124291-25G A124291-5G
ALA124291-100GEA 10303.61 USD
ALA124291-100G ALA124291-10G ALA124291-1G ALA124291250MG ALA124291-25G ALA124291-5G
6-Nitro-o-anisidine ≥97%
6-Nitro-o-anisidine
Formula: C₇H₈N₂O₃
Melting Pt: 73-78°C
MDL Number: MFCD01930197
CAS Number: 16554-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a48981a42bb7af4dc581a609e4401c14d0dbfac&vid=4a48981a42bb7af4dc581a609e4401c14d0dbfac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR