4-Amino-2-bromophenol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Bromo-4-hydroxyaniline

A124279-1G A124279-5G
ALA124279-1GEA 175.66 USD
ALA124279-1G ALA124279-5G
4-Amino-2-bromophenol ≥97%
4-Amino-2-bromophenol
Formula: C₆H₆BrNO
MW: 188.02 g/mol
Melting Pt: 163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06656565
CAS Number: 16750-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0c9cac758ade64966c4adcf7d3046c3756dbfeb&vid=d0c9cac758ade64966c4adcf7d3046c3756dbfeb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR