6-Azauracil ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1,2,4-Triazine-3,5(2H,4H)-dione

A124269-100G A124269-1G A124269-25G A124269-5G
ALA124269-100GEA 684.86 USD
ALA124269-100G ALA124269-1G ALA124269-25G ALA124269-5G
6-Azauracil ≥99%
6-Azauracil

6-Azauracil is used as a growth inhibitor of various microorganisms via depletion of intracellular GTP and UTP nucleotide pools. 6-Azauracil has been widely used in investigations on modulation of transcription, especially in yeast models.A growth inhibitor of various microorganisms

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 274-282°C
MDL Number: MFCD00006456
CAS Number: 461-89-2
EINECS: 207-318-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af08443a4864623cbf3087e8e4ae18eab5a6f63b&vid=af08443a4864623cbf3087e8e4ae18eab5a6f63b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR