8-Azaadenine ≥95%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 3H-[1,2,3]Triazolo[4,5-d]pyrimidin-7-amine

A124265-1G A124265-250MG A124265-5G
ALA124265-1GEA 142.41 USD
ALA124265-1G ALA124265250MG ALA124265-5G
8-Azaadenine ≥95%
8-Azaadenine
Formula: C₄H₄N₆
MW: 136.12 g/mol
Melting Pt: 320°C
Storage Temperature: Refrigerator
MDL Number: MFCD00005697
CAS Number: 1123-54-2
EINECS: 214-375-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=539a103d8260aa91b19768353f80641c60179639&vid=539a103d8260aa91b19768353f80641c60179639&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR