2-Amino-4-fluorophenol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A124264-1G A124264-25G
ALA124264-1GEA 37.91 USD
ALA124264-1G ALA124264-25G
2-Amino-4-fluorophenol ≥97%
2-Amino-4-fluorophenol
Formula: C₆H₆FNO
Melting Pt: 146°C
MDL Number: MFCD00077451
CAS Number: 399-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38776d8a828b09b31a9de3a9ca70f38af54516f7&vid=38776d8a828b09b31a9de3a9ca70f38af54516f7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR