5-Amino-2-chloropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Chloro-5-pyridylamine, 6-Chloropyridin-3-amine, 2-Chloropyridin-5-amine

A124240-1G A124240-25G A124240-5G
ALA124240-1GEA 20.81 USD
ALA124240-1G ALA124240-25G ALA124240-5G
5-Amino-2-chloropyridine ≥98%
5-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 81-84°C
Storage Temperature: Ambient
MDL Number: MFCD00006243
CAS Number: 5350-93-6
EINECS: 226-322-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=673660a9a2ea9170c656fe3eec6dfe67e72b5324&vid=673660a9a2ea9170c656fe3eec6dfe67e72b5324&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR