3-Cyano-4-methylaniline ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Product Application:5-Amino-2-methylbenzonitrile may be used as a starting material for the synthesis of 5-iodo-2-methylbenzonitrile and 5-hydroxy-2-methylbenzonitrile. It may also be used in the preparation of 5-(2,4-dioxothiazolidin-3-yl)-2-methylbenzonitrile by reacting with thioglycolic acid methyl ester in the presence of 1,1′-carbonyldiimidazole.
Formula:
C₈H₈N₂ Boiling Pt: 110 °C Melting Pt: 88-91°C |
MDL Number:
MFCD00017608 CAS Number: 50670-64-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...