2-Aminopyridin-4-ol ≥97% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
2-Amino-4-hydroxypyridine
A124105-1G
A124105-25G
A124105-5G
ALA124105-1GEA
29.36
USD
ALA124105-1G
ALA124105-25G
ALA124105-5G
2-Aminopyridin-4-ol ≥97% (by HPLC)
2-Aminopyridin-4-ol
Formula:
C₅H₆N₂O MW: 110.12 g/mol |
MDL Number:
MFCD04114161 CAS Number: 33631-05-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...