Anthracene-9,10-diboronic acid bis(pinacol) ester ≥97%
Supplier: Aladdin Scientific
|
Warning
|
General description:9,10-Anthracenediboronic acid bis(pinacol) ester is an anthracene based ester that has a conjugated structure. It is an electron rich building block that can be used as an electron donating molecule in the formation of donor-acceptor based organic semiconductor devices. Its planarity and rigid molecular structure allow it to have good charge transporting properties.application:9,10-Anthracenediboronic acid bis(pinacol) ester can be used in the synthesis of polyarylpyrazolines, which can further be utilized in the preparation of organic light emitting diodes (OLEDs).
Formula:
C₂₆H₃₂B₂O₄ Melting Pt: >300°C Storage Temperature: Ambient |
MDL Number:
MFCD16294538 CAS Number: 863992-56-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...