L-Azetidine-2-carboxylic acid ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: L-Trimethyleneimine-2-carboxylic Acid, (S)-Azetidine-2-carboxylic acid, (S)-(-)-2-Azetidinecarboxylic acid, L-Azetidine-2-carboxylic Acid [Antagonist of L-Proline]

A124088-1G A124088-250MG A124088-5G
ALA124088-1GEA 227.91 USD
ALA124088-1G ALA124088250MG ALA124088-5G
L-Azetidine-2-carboxylic acid ≥99%, Moligand™
L-Azetidine-2-carboxylic acid

Product Introduction: L-Azetidine-2-carboxylic acid is a non-protein amino acid and teratogenic agent. It is toxic in nature.Product Application:L-Azetidine-2-carboxylic acid has been used as a: collagen synthesis inhibitor protein folding antagonist as a standard in liquid chromatography-mass spectrometry

Formula: C₄H₇NO₂
MW: 101.11 g/mol
Melting Pt: 217°C
MDL Number: MFCD00005166
CAS Number: 2133-34-8
EINECS: 218-362-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b145a70b28aee131df8c908b6e4d751d9ca0972&vid=9b145a70b28aee131df8c908b6e4d751d9ca0972&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR