2-Acetylthiazole ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(Thiazol-2-yl)-1-ethanone

A124071-100G A124071-25G A124071-5G
ALA124071-100GEA 265.91 USD
ALA124071-100G ALA124071-25G ALA124071-5G
2-Acetylthiazole ≥99%
2-Acetylthiazole

2-Acetylthiazole is useful for preparing triazolothiazoles, chiral alcohols, and in aldol condensation reactions.Useful for preparation of triazolothiazoles,chiral alcohols,and in aldol condensation reactions.

Formula: C₅H₅NOS
MW: 127.17 g/mol
Boiling Pt: 90°C
Density: 1.227 g/cm³
Flash Pt: 78°C
Storage Temperature: Ambient
MDL Number: MFCD00005324
CAS Number: 24295-03-2
EINECS: 246-134-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=613e4d2700f0595e548079f9d3675d131cd4be69&vid=613e4d2700f0595e548079f9d3675d131cd4be69&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR