N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-(3-pyridyl)-2-pyrimidineamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine, N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine

A124052-100G A124052-1G A124052-25G A124052-5G
ALA124052-100GEA 3934.81 USD
ALA124052-100G ALA124052-1G ALA124052-25G ALA124052-5G
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine ≥98%
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine

N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine is a selective inhibitor of protein kinase C (PKC).

Formula: C₁₆H₁₅N₅
Melting Pt: 145 °C
MDL Number: MFCD09028125
CAS Number: 152460-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72135f1fee97d1d5b12a01051a496c59907274dd&vid=72135f1fee97d1d5b12a01051a496c59907274dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR