4-Amino-3-nitrobenzonitrile ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Cyano-2-nitroaniline

A124035-1G A124035-5G
ALA124035-1GEA 21.76 USD
ALA124035-1G ALA124035-5G
4-Amino-3-nitrobenzonitrile ≥98% (by GC)
4-Amino-3-nitrobenzonitrile
Formula: C₇H₅N₃O₂
Melting Pt: 160.0-164.0°C
MDL Number: MFCD00013373
CAS Number: 6393-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c69b80e23d7d4627baaa9ff1ce011e867de1ae5&vid=0c69b80e23d7d4627baaa9ff1ce011e867de1ae5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR