4-Amino-3-chloro-5-methylbenzonitrile ≥90%

Supplier: ALADDIN SCIENTIFIC
Warning

A124034-1G A124034-25G A124034-5G
ALA124034-1GEA 73.06 USD
ALA124034-1G ALA124034-25G ALA124034-5G
4-Amino-3-chloro-5-methylbenzonitrile ≥90%
4-Amino-3-chloro-5-methylbenzonitrile
Formula: C₈H₇ClN₂
MW: 166.61 g/mol
Melting Pt: 128-130°C
MDL Number: MFCD00041476
CAS Number: 158296-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77cba7480f6eb7eaa23861b77475cf9ce5a3de51&vid=77cba7480f6eb7eaa23861b77475cf9ce5a3de51&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR