Ammonium tetrachloroaurate hydrate ≥99.99% (metals basis), PrimorTrace™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Ammonium tetrachloroaurate(III) hydrate

A124024-1G A124024-5G
ALA124024-1GEA 123.41 USD
ALA124024-1G ALA124024-5G
Ammonium tetrachloroaurate hydrate ≥99.99% (metals basis), PrimorTrace™
Ammonium tetrachloroaurate hydrate
Formula: H₄AuCl₄N·H₂O
MW: 356.82 g/mol (anhy)
MDL Number: MFCD00011503
CAS Number: 13874-04-9
EINECS: 250-476-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e5d3d6e6ee7099895118c6c56e96bdf0e228d7a&vid=6e5d3d6e6ee7099895118c6c56e96bdf0e228d7a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR