Ammonium hexachlororuthenate(IV) Ru 28.4% min

Supplier: ALADDIN SCIENTIFIC
Danger

A124007-100MG A124007-1G A124007-200MG
ALA124007100MGEA 12.26 USD
ALA124007100MG ALA124007-1G ALA124007200MG
Ammonium hexachlororuthenate(IV) Ru 28.4% min
Ammonium hexachlororuthenate(IV)
Formula: H₈Cl₆N₂Ru
Storage Temperature: Ambient
MDL Number: MFCD00015962
CAS Number: 18746-63-9
EINECS: 242-552-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81822b5e92727a1fb7a4274cad337fb3f18698c6&vid=81822b5e92727a1fb7a4274cad337fb3f18698c6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR