2-Amino-6-methylbenzonitrile ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
2-Cyano-3-methylaniline
A123972-10G
A123972-1G
A123972-250MG
A123972-25G
A123972-5G
ALA123972-10GEA
221.26
USD
ALA123972-10G
ALA123972-1G
ALA123972250MG
ALA123972-25G
ALA123972-5G
2-Amino-6-methylbenzonitrile ≥97%
2-Amino-6-methylbenzonitrile
may discolor to brown on storage
Formula:
C₈H₈N₂ MW: 132.16 g/mol Melting Pt: 127-132°C Storage Temperature: Ambient |
MDL Number:
MFCD00058686 CAS Number: 56043-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...