2-Amino-5-fluorobenzonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A123971-1G A123971-5G
ALA123971-1GEA 36.96 USD
ALA123971-1G ALA123971-5G
2-Amino-5-fluorobenzonitrile ≥97%
2-Amino-5-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Boiling Pt: 138°C
Melting Pt: 92-96°C
MDL Number: MFCD02683869
CAS Number: 61272-77-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c238564cdadcb4dc2a6e5d9060b19f104d08e533&vid=c238564cdadcb4dc2a6e5d9060b19f104d08e533&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR