2-Amino-4-methylbenzonitrile ≥98% (by GC)
Supplier: Aladdin Scientific
|
Warning
|
A123969-1G
A123969-25G
A123969-5G
ALA123969-1GEA
29.36
USD
ALA123969-1G
ALA123969-25G
ALA123969-5G
2-Amino-4-methylbenzonitrile ≥98% (by GC)
2-Amino-4-methylbenzonitrile
It was used in the synthesis of: 7-methyl-4-(phenylamino)quinazoline-2(1H)-selone. racemic aminoquinolines, potential acetylcholinesterase (AChE) inhibitors.
Formula:
C₈H₈N₂ Melting Pt: 93.0-97.0°C Storage Temperature: Ambient |
MDL Number:
MFCD00173706 CAS Number: 26830-96-6 EINECS: 248-020-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...