2-Amino-3-chlorobenzonitrile ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A123967-1G
A123967-5G
ALA123967-1GEA
109.16
USD
ALA123967-1G
ALA123967-5G
2-Amino-3-chlorobenzonitrile ≥97%
2-Amino-3-chlorobenzonitrile
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol Melting Pt: 94-96°C |
MDL Number:
MFCD03425881 CAS Number: 53312-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...