(S)-(-)-3-Aminoquinuclidine dihydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (S)-Quinuclidin-3-amine dihydrochloride, (S)-Quinuclidin-3-amine 2HCl, (S)-(-)-3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

A123962-1G A123962-250MG A123962-25G A123962-5G
ALA123962-1GEA 28.41 USD
ALA123962-1G ALA123962250MG ALA123962-25G ALA123962-5G
(S)-(-)-3-Aminoquinuclidine dihydrochloride ≥98%
(S)-(-)-3-Aminoquinuclidine dihydrochloride
Formula: C₇H₁₆Cl₂N₂
MW: 199.1213 g/mol
MDL Number: MFCD00191753
CAS Number: 119904-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e953f0e4abfab539ecea1f3b2d52ad399a444295&vid=e953f0e4abfab539ecea1f3b2d52ad399a444295&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR