5-Amino-2-bromobenzoic acid ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A123936-1G A123936-25G A123936-5G
ALA123936-1GEA 14.16 USD
ALA123936-1G ALA123936-25G ALA123936-5G
5-Amino-2-bromobenzoic acid ≥95%
5-Amino-2-bromobenzoic acid
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 366.2°C
Melting Pt: 171-179°C
Density: 1.79 g/cm³
MDL Number: MFCD07368968
CAS Number: 2840-02-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5feafab1c40f77807a7c12995ed6174cd5ef3989&vid=5feafab1c40f77807a7c12995ed6174cd5ef3989&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR