3-Amino-4-iodopyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A123932-1G A123932-5G
ALA123932-1GEA 118.66 USD
ALA123932-1G ALA123932-5G
3-Amino-4-iodopyridine ≥98%
3-Amino-4-iodopyridine
Formula: C₅H₅IN₂
Melting Pt: 55-70°C
Storage Temperature: Ambient
MDL Number: MFCD04971334
CAS Number: 105752-11-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c214e4528cb0a226b9b77694d8299569677949b&vid=9c214e4528cb0a226b9b77694d8299569677949b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR