4-Amino-3-chlorobenzoic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A123912-100G A123912-1G A123912-25G A123912-5G
ALA123912-100GEA 777.96 USD
ALA123912-100G ALA123912-1G ALA123912-25G ALA123912-5G
4-Amino-3-chlorobenzoic acid ≥97%
4-Amino-3-chlorobenzoic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 224-229°C
Storage Temperature: Ambient
MDL Number: MFCD00460423
CAS Number: 2486-71-7
EINECS: 219-630-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa4a895c5a1b7142b7305b81dd2b26c4023ce5d6&vid=aa4a895c5a1b7142b7305b81dd2b26c4023ce5d6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR