3-Amino-2,6-dichloropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2,6-Dichloro-3-pyridinamine, 2,6-Dichloropyridin-3-amine, 2,6-Dichloro-3-pyridylamine

A123903-1G A123903-25G A123903-5G
ALA123903-1GEA 12.26 USD
ALA123903-1G ALA123903-25G ALA123903-5G
3-Amino-2,6-dichloropyridine ≥98%
3-Amino-2,6-dichloropyridine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Melting Pt: 120-124°C
MDL Number: MFCD00023417
CAS Number: 62476-56-6
EINECS: 263-559-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4be25c87f545843a5229780bf4fe6b1bad138b72&vid=4be25c87f545843a5229780bf4fe6b1bad138b72&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR