2-Amino-3-iodopyridine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A123901-25G
A123901-5G
A123901-1G
ALA123901-25GEA
511.96
USD
ALA123901-25G
ALA123901-5G
ALA123901-1G
2-Amino-3-iodopyridine ≥98%
2-Amino-3-iodopyridine
Formula:
C₅H₅IN₂ Melting Pt: 88-92°C |
MDL Number:
MFCD01651885 CAS Number: 104830-06-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...