3-Amino-2-chlorobenzoic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A123890-1G A123890-5G
ALA123890-1GEA 51.21 USD
ALA123890-1G ALA123890-5G
3-Amino-2-chlorobenzoic acid ≥97%
3-Amino-2-chlorobenzoic acid
Formula: C7H6ClNO2
MW: 171.5810 g/mol
Melting Pt: 154-160°C
MDL Number: MFCD00191726
CAS Number: 108679-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86a82ed6dcaaa0c0dc9a4045b7cf3ade172a16bc&vid=86a82ed6dcaaa0c0dc9a4045b7cf3ade172a16bc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR