5-Methoxyanthranilic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A123845-100G A123845-1G A123845-25G A123845-5G
ALA123845-100GEA 2164.01 USD
ALA123845-100G ALA123845-1G ALA123845-25G ALA123845-5G
5-Methoxyanthranilic acid ≥98%
5-Methoxyanthranilic acid

Application:2-Amino-5-methoxybenzoic acid is a general reagent used in the synthesis of substituted isoquinolinonaphthyridines, quinazolinones, imidazobenzodiazepines, pyridoquinazolones and polycyclic hexahydrobenzo[c]acridines.Product Introduction:2-Amino-5-methoxybenzoic Acid is used in the synthesis of quinazolinone class of histamine H3 receptor inverse agonists. It is also used in the synthesis of Alogliptin, a selective inhibitor of the serine protease dipeptidyl peptidase IV.

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 148-152°C
MDL Number: MFCD00016509
CAS Number: 6705-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR