4-Methoxyanthranilic acid ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
2-Amino-4-methoxybenzoic acid
A123844-1G
A123844-25G
A123844-5G
ALA123844-1GEA
43.61
USD
ALA123844-1G
ALA123844-25G
ALA123844-5G
4-Methoxyanthranilic acid ≥98%
4-Methoxyanthranilic acid
Formula:
C₈H₉NO₃ MW: 167.16 g/mol Melting Pt: 175-180°C |
MDL Number:
MFCD00667729 CAS Number: 4294-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...