(3R)-3-Aminobutan-1-ol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 1-Butanol,3-aMino-, (3R)-, (R)-3-Aminobutanol, (R)-3-Amino-1-butanol, (R)-3-Aminobutan-1-ol, (-)-(R)-3-aMinopropan-1-ol, (3R)-3-Amino-1-butanol, R-3-amino-1-butanol

A123608-100G A123608-1G A123608-25G A123608-500G A123608-5G
ALA123608-100GEA 4824.96 USD
ALA123608-100G ALA123608-1G ALA123608-25G ALA123608-500G ALA123608-5G
(3R)-3-Aminobutan-1-ol ≥98%
(3R)-3-Aminobutan-1-ol
Formula: C₄H₁₁NO
MW: 89.1362 g/mol
Boiling Pt: 168°C
Density: 0.927 g/cm³
Flash Pt: 56 °C
MDL Number: MFCD13184351
CAS Number: 61477-40-5
UN: 2734
ADR: 8(3),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09dffbe5f728ce931035a127e0eafed5e096547f&vid=09dffbe5f728ce931035a127e0eafed5e096547f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR