2-Amino-4,6-dihydroxypyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-6-hydroxy-1H-pyrimidin-4-one, 2-Amino-6-hydroxypyrimidin-4(3H)-one

A123505-100G A123505-25G
ALA123505-100GEA 127.21 USD
ALA123505-100G ALA123505-25G
2-Amino-4,6-dihydroxypyrimidine ≥98%
2-Amino-4,6-dihydroxypyrimidine
Formula: C₄H₅N₃O₂
MW: 127.1 g/mol
Melting Pt: >300°C
Storage Temperature: Refrigerator
MDL Number: MFCD00006094
CAS Number: 56-09-7
EINECS: 200-256-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR