2-Amino-3-thiophenecarbonitrile ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-Aminothiophene-3-carbonitrile

A123466-1G A123466-25G A123466-5G
ALA123466-1GEA 14.16 USD
ALA123466-1G ALA123466-25G ALA123466-5G
2-Amino-3-thiophenecarbonitrile ≥97% (by GC)
2-Amino-3-thiophenecarbonitrile
Formula: C₅H₄N₂S
MW: 124.16 g/mol
Melting Pt: 104-108°C
Storage Temperature: Ambient
MDL Number: MFCD00706298
CAS Number: 4651-82-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f5c2ae78b34631c150697bb7b251599c46c6cb3&vid=2f5c2ae78b34631c150697bb7b251599c46c6cb3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR