2-Amino-4-bromophenol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 5-Bromo-2-hydroxyaniline

A123465-250MG A123465-25G A123465-5G
ALA123465250MGEA 9.41 USD
ALA123465250MG ALA123465-25G ALA123465-5G
2-Amino-4-bromophenol ≥98%
2-Amino-4-bromophenol

Reactant for: 1.Preparation of (aryl)oxadiazolobenzoxazinones via Suzuki-Miyaura reaction. 2.Copper-catalyzed oxidative amination of benzoxazoles and related azoles via C-H and C-N bond activation. 3.Synthesis of N-(alkoxyphenyl)-aminocarbonylbenzoic acid derivatives as protein tyrosine phophatase 1B inhibitors. 4. Preparation of lipophilic deoxy-xylulose-phosphate reductoisomerase inhibitors. 5.Synthesis of benzoxazole benzenesulfonamides as allosteric inhibitors of fructose-1,6-bisphosphatase.application:Reactant for:Preparation of (aryl)oxadiazolobenzoxazinones via Suzuki-Miyaura reactionCopper-catalyzed oxidative amination of benzoxazoles and related azoles via C-H and C-N bond activationSynthesis of N-(alkoxyphenyl)-aminocarbonylbenzoic acid derivatives as protein tyrosine phophatase 1B inhibitorsPreparation of lipophilic deoxy-xylulose-phosphate reductoisomerase inhibitorsSynthesis of benzoxazole benzenesulfonamides as allosteric inhibitors of fructose-1,6-bisphosphatase

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Melting Pt: 133°C
Storage Temperature: Refrigerator
MDL Number: MFCD00235171
CAS Number: 40925-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae489cc1cf6ccff5a70c3102c51f8db40b7d6c68&vid=ae489cc1cf6ccff5a70c3102c51f8db40b7d6c68&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR