5-Amino-2,4-dichloropyrimidine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2,4-Dichloropyrimidin-5-amine

A123458-1G A123458-250MG
ALA123458-1GEA 63.56 USD
ALA123458-1G ALA123458250MG
5-Amino-2,4-dichloropyrimidine ≥97%
5-Amino-2,4-dichloropyrimidine

A pyrimidinamine derivative which may act as a Plk1 inhibitor

Formula: C₄H₃Cl₂N₃
MW: 164.00 g/mol
Melting Pt: 118-123°C
Storage Temperature: Refrigerator
MDL Number: MFCD05662684
CAS Number: 5177-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2029975ff72d8a5939d0e407d21db67bc712999&vid=d2029975ff72d8a5939d0e407d21db67bc712999&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR