2-Aminonicotinamide ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-Aminonicotinamide, 2-Aminopyridine-3-carboxamide

A123442-1G A123442-5G
ALA123442-1GEA 45.51 USD
ALA123442-1G ALA123442-5G
2-Aminonicotinamide ≥97%
2-Aminonicotinamide
Formula: C₆H₇N₃O
MW: 137.142 g/mol
Melting Pt: 194-200°C
MDL Number: MFCD00128873
CAS Number: 13438-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d027b4adaa727aa82127f8cfed6ab78da3b2c5c5&vid=d027b4adaa727aa82127f8cfed6ab78da3b2c5c5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR