2-Amino-4,6-dimethylpyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A123441-25G A123441-5G
ALA123441-25GEA 122.46 USD
ALA123441-25G ALA123441-5G
2-Amino-4,6-dimethylpyridine ≥98%
2-Amino-4,6-dimethylpyridine
Formula: C₇H₁₀N₂
Boiling Pt: 231°C
Melting Pt: 67°C
MDL Number: MFCD00006322
CAS Number: 5407-87-4
EINECS: 226-470-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1e534cd4fba07ee2b59918748e715be4d99b8d0&vid=d1e534cd4fba07ee2b59918748e715be4d99b8d0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR