2-Amino-6-chloropyrazine ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A123436-100G
A123436-1G
ALA123436-100GEA
1097.16
USD
ALA123436-100G
ALA123436-1G
2-Amino-6-chloropyrazine ≥98% (by GC)
2-Amino-6-chloropyrazine
Application:2-Amino-6-chloropyrazine is used in the preparation of A2B adenosine receptor antagonists and other biologically active compounds.
Formula:
C₄H₄ClN₃ Melting Pt: 153°C |
MDL Number:
MFCD00055024 CAS Number: 33332-28-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...