2-Amino-5-bromopyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Bromopyrimidin-2-ylamine, 5-Bromopyrimidin-2-amine

A123409-100G A123409-1G A123409-25G A123409-5G
ALA123409-100GEA 523.36 USD
ALA123409-100G ALA123409-1G ALA123409-25G ALA123409-5G
2-Amino-5-bromopyrimidine ≥98%
2-Amino-5-bromopyrimidine

Intermediate for the preparation of sulfanilamides and amino acids containing the pyrimidine ring system. The products are potential antiviral agents.application:2-Amino-5-bromopyrimidine was employed:in synthesis of pyridine, pyrimidine and pyridinone C-nucleoside phosphoramiditesin synthesis of 2-phthalimido-5-bromopyrimidineas intermediate for the preparation of sulfanilamides and amino acids containing the pyrimidine ring system. The products are potential antiviral agents.

Formula: C₄H₄BrN₃
MW: 174 g/mol
Melting Pt: 244°C
Storage Temperature: Refrigerator
MDL Number: MFCD00012341
CAS Number: 7752-82-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95de0037999ae66a093b7366503bb73b5b656fdf&vid=95de0037999ae66a093b7366503bb73b5b656fdf&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR