2-Allyl-4,6-dibenzoylresorcinol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: (5-Allyl-4,6-dihydroxy-1,3-phenylene)bis(phenylmethanone)

A123367-100G A123367-25G A123367-5G
ALA123367-100GEA 868.21 USD
ALA123367-100G ALA123367-25G ALA123367-5G
2-Allyl-4,6-dibenzoylresorcinol ≥98%
2-Allyl-4,6-dibenzoylresorcinol
Formula: C₂₃H₁₈O₄
MW: 358.40 g/mol
Melting Pt: 158-163°C
MDL Number: MFCD02094038
CAS Number: 102593-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac9c5cd6fbb6941eb51a09100b4432198597d776&vid=ac9c5cd6fbb6941eb51a09100b4432198597d776&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR