2-Amino-4-ethylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Ethylpyridin-2-amine, 4-Ethyl-2-pyridylamine

A123353-10G A123353-1G A123353-2.5G
ALA123353-10GEA 183.26 USD
ALA123353-10G ALA123353-1G ALA123353-2.5G
2-Amino-4-ethylpyridine ≥97%
2-Amino-4-ethylpyridine

Soluble in Hexane, Ethyl Acetate and Methyl tert-Butyl Ether

Formula: C₇H₁₀N₂
Melting Pt: 65-69°C
MDL Number: MFCD00129025
CAS Number: 33252-32-3
EINECS: 251-430-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4e11d2a23ec5d149e0f3dc448e43c8f25a54f1c&vid=b4e11d2a23ec5d149e0f3dc448e43c8f25a54f1c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR